3-Chloro-4-ethoxy-2-(trimethylsilyl)benzoic acid ≥97%

Supplier: AOBChem USA

Warning

73652-500MG 73652-1G 73652-5G 73652-25G
77333-934EA 136.99 USD
77333-934 77344-636 77369-944 77390-812
3-Chloro-4-ethoxy-2-(trimethylsilyl)benzoic acid ≥97%
3-Chloro-4-ethoxy-2-(trimethylsilyl)benzoic acid

Precaución: for Research Use Only

Formula: C₁₂H₁₇ClO₃Si
MW: 272.8 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34765092
CAS Number: 2901103-05-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR