4-Fluoro-3-methoxy-5-(trimethylsilyl)benzoic acid ≥97%

Supplier: AOBChem USA

Warning

73631-500MG 73631-1G 73631-5G 73631-25G
77333-932EA 203.94 USD
77333-932 77358-092 77369-942 77390-810
4-Fluoro-3-methoxy-5-(trimethylsilyl)benzoic acid ≥97%
4-Fluoro-3-methoxy-5-(trimethylsilyl)benzoic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₅FO₃Si
MW: 242.319 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34765075
CAS Number: 2901065-37-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR