3,5-Dimethoxy-4-(methoxymethyl)benzoic acid ≥97%

Supplier: AOBChem USA

Warning

73587-500MG 73587-1G 73587-5G
77339-994EA 706.58 USD
77339-994 77358-082 77382-002
3,5-Dimethoxy-4-(methoxymethyl)benzoic acid ≥97%
3,5-Dimethoxy-4-(methoxymethyl)benzoic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₄O₅
MW: 226.226 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34765042
CAS Number: 1282549-93-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR