1,2-Dibromo-4-methoxy-5-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

73561-500MG 73561-1G 73561-5G 73561-25G
77333-926EA 141.11 USD
77333-926 77344-928 77369-938 77390-806
1,2-Dibromo-4-methoxy-5-methylbenzene ≥97%
1,2-Dibromo-4-methoxy-5-methylbenzene

Precaución: for Research Use Only

Formula: C₈H₈Br₂O
Storage Temperature: Ambient
MDL Number: MFCD28053985
CAS Number: 1781035-42-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR