2-Iodo-4-isopropyl-1-methoxybenzene ≥95%

Supplier: AOBChem USA

Warning

73530-500MG 73530-1G 73530-5G
77335-726EA 363.59 USD
77335-726 77358-080 77381-998
2-Iodo-4-isopropyl-1-methoxybenzene ≥95%
2-Iodo-4-isopropyl-1-methoxybenzene

Precaución: for Research Use Only

Formula: C₁₀H₁₃IO
MW: 276.114 g/mol
Storage Temperature: Ambient
MDL Number: MFCD29096398
CAS Number: 1369897-14-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR