(5-Isopropyl-2-methoxyphenyl)(methyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

73529-500MG 73529-1G 73529-5G
77338-232EA 469.68 USD
77338-232 77358-078 77381-996
(5-Isopropyl-2-methoxyphenyl)(methyl)sulfane ≥97%
(5-Isopropyl-2-methoxyphenyl)(methyl)sulfane

Precaución: for Research Use Only

Formula: C₁₁H₁₆OS
MW: 196.309 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34765002
CAS Number: 2807465-67-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR