1-(5-Isopropyl-2-methoxyphenyl)ethanone ≥95%

Supplier: AOBChem USA

Warning

73527-500MG 73527-1G 73527-5G 73527-25G
77333-922EA 161.71 USD
77333-922 77345-998 77369-934 77390-804
1-(5-Isopropyl-2-methoxyphenyl)ethanone ≥95%
1-(5-Isopropyl-2-methoxyphenyl)ethanone

Precaución: for Research Use Only

Formula: C₁₂H₁₆O₂
Storage Temperature: Ambient
MDL Number: MFCD10485788
CAS Number: 830329-27-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR