[2,3'-Bipyridine]-5-carboxylic acid ≥97%

Supplier: AOBChem USA

Warning

73470-500MG 73470-1G 73470-5G
77333-918EA 307.97 USD
77333-918 77358-076 77381-992
[2,3'-Bipyridine]-5-carboxylic acid ≥97%
[2,3'-Bipyridine]-5-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₁H₈N₂O₂
Storage Temperature: Ambient
MDL Number: MFCD15476024
CAS Number: 5059-52-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR