3-Acetyl-5-methylbenzoic acid ≥97%

Supplier: AOBChem USA

Warning

73442-500MG 73442-1G 73442-25G 73442-5G
77333-916EA 297.67 USD
77333-916 77358-074 77390-802 77381-990
3-Acetyl-5-methylbenzoic acid ≥97%
3-Acetyl-5-methylbenzoic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₀O₃
MW: 178.185 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22544378
CAS Number: 1393540-28-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR