(2,4-Difluoro-3-iodophenyl)methanol ≥97%

Supplier: AOBChem USA

Warning

73440-500MG 73440-1G 73440-5G
77333-914EA 344.02 USD
77333-914 77358-072 77381-988
(2,4-Difluoro-3-iodophenyl)methanol ≥97%
(2,4-Difluoro-3-iodophenyl)methanol

Precaución: for Research Use Only

Formula: C₇H₅F₂IO
MW: 270.015 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34764967
CAS Number: 1628040-36-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR