1,4-Dichloro-2-fluoro-5-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

73375-500MG 73375-1G 73375-5G
77333-912EA 338.87 USD
77333-912 77358-068 77381-984
1,4-Dichloro-2-fluoro-5-methylbenzene ≥97%
1,4-Dichloro-2-fluoro-5-methylbenzene

Precaución: for Research Use Only

Formula: C₇H₅Cl₂F
MW: 179.019 g/mol
Storage Temperature: Ambient
MDL Number: MFCD21602119
CAS Number: 1365969-73-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR