1,5-Dichloro-2-methyl-4-(methylsulfonyl)benzene ≥97%

Supplier: AOBChem USA

Warning

73358-500MG 73358-1G 73358-5G
77338-676EA 507.79 USD
77338-676 77358-066 77381-982
1,5-Dichloro-2-methyl-4-(methylsulfonyl)benzene ≥97%
1,5-Dichloro-2-methyl-4-(methylsulfonyl)benzene

Precaución: for Research Use Only

Formula: C₈H₈Cl₂O₂S
MW: 239.119 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34764908
CAS Number: 2807448-94-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR