1-(5-Fluoro-4-methoxy-2-methylphenyl)ethanol ≥97%

Supplier: AOBChem USA

Warning

73343-500MG 73343-1G 73343-5G 73343-25G
77333-908EA 282.22 USD
77333-908 77358-062 77369-928 77390-798
1-(5-Fluoro-4-methoxy-2-methylphenyl)ethanol ≥97%
1-(5-Fluoro-4-methoxy-2-methylphenyl)ethanol

Precaución: for Research Use Only

Formula: C₁₀H₁₃FO₂
Storage Temperature: Ambient
MDL Number: MFCD34764898
CAS Number: 2807446-17-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR