2,2,2-Trifluoro-1-(5-fluoro-4-methoxy-2-methylphenyl)ethanone ≥97%

Supplier: AOBChem USA

Warning

73340-500MG 73340-1G 73340-5G
77335-652EA 361.53 USD
77335-652 77358-060 77381-980
2,2,2-Trifluoro-1-(5-fluoro-4-methoxy-2-methylphenyl)ethanone ≥97%
2,2,2-Trifluoro-1-(5-fluoro-4-methoxy-2-methylphenyl)ethanone

Precaución: for Research Use Only

Formula: C₁₀H₈F₄O₂
MW: 236.163 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34764896
CAS Number: 2901103-01-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR