2-(Benzyloxy)-4-bromo-6-methylaniline ≥97%

Supplier: AOBChem USA

Warning

73250-500MG 73250-1G 73250-5G 73250-25G
77333-904EA 252.35 USD
77333-904 77358-058 77369-924 77390-796
2-(Benzyloxy)-4-bromo-6-methylaniline ≥97%
2-(Benzyloxy)-4-bromo-6-methylaniline

Precaución: for Research Use Only

Formula: C₁₄H₁₄BrNO
MW: 292.171 g/mol
Storage Temperature: Ambient
MDL Number: MFCD26398838
CAS Number: 928014-67-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR