2-(Benzyloxy)-6-methylaniline ≥97%

Supplier: AOBChem USA

Warning

73249-500MG 73249-1G 73249-5G 73249-25G
77333-902EA 161.71 USD
77333-902 77345-996 77369-922 77390-794
2-(Benzyloxy)-6-methylaniline ≥97%
2-(Benzyloxy)-6-methylaniline

Precaución: for Research Use Only

Formula: C₁₄H₁₅NO
Storage Temperature: Ambient
MDL Number: MFCD06410774
CAS Number: 883107-58-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR