3-Isopropoxy-2-nitrobenzoic acid ≥97%

Supplier: AOBChem USA

Warning

73247-500MG 73247-1G 73247-5G 73247-25G
77333-900EA 202.91 USD
77333-900 77358-056 77369-920 77390-792
3-Isopropoxy-2-nitrobenzoic acid ≥97%
3-Isopropoxy-2-nitrobenzoic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₁NO₅
MW: 225.198 g/mol
Storage Temperature: Ambient
MDL Number: MFCD24227687
CAS Number: 1369928-45-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR