4-Bromo-2-isopropoxy-6-methylaniline ≥97%

Supplier: AOBChem USA

Warning

73246-500MG 73246-1G 73246-5G 73246-25G
77333-898EA 202.91 USD
77333-898 77358-054 77369-918 77390-790
4-Bromo-2-isopropoxy-6-methylaniline ≥97%
4-Bromo-2-isopropoxy-6-methylaniline

Precaución: for Research Use Only

Formula: C₁₀H₁₄BrNO
MW: 244.128 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34764841
CAS Number: 2807441-87-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR