2-Isopropoxy-6-methylaniline ≥95%

Supplier: AOBChem USA

Warning

73245-500MG 73245-1G 73245-5G 73245-25G
77333-896EA 157.59 USD
77333-896 77345-722 77369-916 77390-788
2-Isopropoxy-6-methylaniline ≥95%
2-Isopropoxy-6-methylaniline

Precaución: for Research Use Only

Formula: C₁₀H₁₅NO
Storage Temperature: Ambient
MDL Number: MFCD12170085
CAS Number: 187471-36-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR