4-Borono-2-fluoro-3-methoxybenzoic acid ≥97%

Supplier: AOBChem USA

Warning

73211-500MG 73211-1G 73211-5G
77335-650EA 361.53 USD
77335-650 77358-052 77381-976
4-Borono-2-fluoro-3-methoxybenzoic acid ≥97%
4-Borono-2-fluoro-3-methoxybenzoic acid

Precaución: for Research Use Only

Formula: C₈H₈BFO₅
MW: 213.956 g/mol
Storage Temperature: Ambient
MDL Number: MFCD31636826
CAS Number: 1692914-76-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR