1-Iodo-3-propoxy-5-(trifluoromethoxy)benzene ≥97%

Supplier: AOBChem USA

Warning

73177-500MG 73177-1G 73177-5G 73177-25G
77333-888EA 184.37 USD
77333-888 77347-338 77369-912 77390-782
1-Iodo-3-propoxy-5-(trifluoromethoxy)benzene ≥97%
1-Iodo-3-propoxy-5-(trifluoromethoxy)benzene

Precaución: for Research Use Only

Formula: C₁₀H₁₀F₃IO₂
MW: 346.085 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34764786
CAS Number: 2807455-07-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR