(3-Chloro-2-isopropoxy-4-methylphenyl)(methyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

73170-500MG 73170-1G 73170-5G 73170-25G
77333-886EA 190.55 USD
77333-886 77358-044 77369-910 77390-780
(3-Chloro-2-isopropoxy-4-methylphenyl)(methyl)sulfane ≥97%
(3-Chloro-2-isopropoxy-4-methylphenyl)(methyl)sulfane

Precaución: for Research Use Only

Formula: C₁₁H₁₅ClOS
Storage Temperature: Ambient
MDL Number: MFCD34764780
CAS Number: 2807470-23-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR