1-Bromo-3-chloro-2-isopropoxy-4-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

73158-500MG 73158-1G 73158-5G 73158-25G
77333-884EA 90.64 USD
77333-884 77342-404 77369-908 77381-970
1-Bromo-3-chloro-2-isopropoxy-4-methylbenzene ≥97%
1-Bromo-3-chloro-2-isopropoxy-4-methylbenzene

Precaución: for Research Use Only

Formula: C₁₀H₁₂BrClO
MW: 263.559 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34764768
CAS Number: 2807470-82-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR