4-Chloro-3-fluoro-5-methylbenzoic acid ≥97%

Supplier: AOBChem USA

Warning

73118-500MG 73118-1G 73118-5G
77339-090EA 548.99 USD
77339-090 77358-038 77381-964
4-Chloro-3-fluoro-5-methylbenzoic acid ≥97%
4-Chloro-3-fluoro-5-methylbenzoic acid

Precaución: for Research Use Only

Formula: C₈H₆ClFO₂
MW: 188.583 g/mol
Storage Temperature: Ambient
MDL Number: MFCD23712097
CAS Number: 1427418-68-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR