1-Bromo-4-(methylsulfonyl)naphthalene ≥97%

Supplier: AOBChem USA

Warning

73085-500MG 73085-1G 73085-5G 73085-25G
77333-878EA 182.31 USD
77333-878 77347-030 77369-904 77390-776
1-Bromo-4-(methylsulfonyl)naphthalene ≥97%
1-Bromo-4-(methylsulfonyl)naphthalene

Precaución: for Research Use Only

Formula: C₁₁H₉BrO₂S
Storage Temperature: Ambient
MDL Number: MFCD10081722
CAS Number: 63279-40-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR