4-(6-Chloro-5-methylpyridin-3-yl)benzaldehyde ≥97%

Supplier: AOBChem USA

Warning

73059-500MG 73059-1G 73059-5G 73059-25G
77333-872EA 83.43 USD
77333-872 77342-182 77369-898 77381-958
4-(6-Chloro-5-methylpyridin-3-yl)benzaldehyde ≥97%
4-(6-Chloro-5-methylpyridin-3-yl)benzaldehyde

Precaución: for Research Use Only

Formula: C₁₃H₁₀ClNO
MW: 231.678 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34764706
CAS Number: 2901098-67-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR