3-(6-Chloro-5-methylpyridin-3-yl)benzaldehyde ≥97%

Supplier: AOBChem USA

Warning

73058-1G 73058-5G 73058-25G
77341-812EA 104.03 USD
77341-812 77369-896 77381-956
3-(6-Chloro-5-methylpyridin-3-yl)benzaldehyde ≥97%
3-(6-Chloro-5-methylpyridin-3-yl)benzaldehyde

Precaución: for Research Use Only

Formula: C₁₃H₁₀ClNO
MW: 231.678 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34764705
CAS Number: 2901105-07-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR