(2,6-Dichloro-3-methylphenyl)(morpholino)methanone ≥97%

Supplier: AOBChem USA

Warning

73052-500MG 73052-1G 73052-5G 73052-25G
77333-870EA 136.99 USD
77333-870 77344-566 77369-894 77390-772
(2,6-Dichloro-3-methylphenyl)(morpholino)methanone ≥97%
(2,6-Dichloro-3-methylphenyl)(morpholino)methanone

Precaución: for Research Use Only

Formula: C₁₂H₁₃Cl₂NO₂
MW: 274.143 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34764701
CAS Number: 2918865-51-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR