2,6-Dichloro-3-methylbenzamide ≥97%

Supplier: AOBChem USA

Warning

73043-500MG 73043-1G 73043-5G 73043-25G
77333-864EA 169.95 USD
77333-864 77346-370 77369-888 77390-766
2,6-Dichloro-3-methylbenzamide ≥97%
2,6-Dichloro-3-methylbenzamide

Precaución: for Research Use Only

Formula: C₈H₇Cl₂NO
Storage Temperature: Ambient
MDL Number: MFCD28786760
CAS Number: 1803835-81-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR