1,5-Dichloro-2-(methoxymethyl)-4-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

73038-500MG 73038-1G 73038-5G
77337-336EA 410.97 USD
77337-336 77358-028 77381-954
1,5-Dichloro-2-(methoxymethyl)-4-methylbenzene ≥97%
1,5-Dichloro-2-(methoxymethyl)-4-methylbenzene

Precaución: for Research Use Only

Formula: C₉H₁₀Cl₂O
MW: 205.081 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34764692
CAS Number: 2901082-09-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR