5-Bromo-2,4-dichloro-N-methoxy-N-methylbenzamide ≥97%

Supplier: AOBChem USA

Warning

73032-500MG 73032-1G 73032-5G 73032-25G
77333-862EA 121.54 USD
77333-862 77343-878 77369-886 77381-952
5-Bromo-2,4-dichloro-N-methoxy-N-methylbenzamide ≥97%
5-Bromo-2,4-dichloro-N-methoxy-N-methylbenzamide

Precaución: for Research Use Only

Formula: C₉H₈BrCl₂NO₂
Storage Temperature: Ambient
MDL Number: MFCD34764686
CAS Number: 2901085-64-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR