(2,4-Dichloro-5-(methylthio)phenyl)methanol ≥97%

Supplier: AOBChem USA

Warning

73020-500MG 73020-1G 73020-5G
77337-644EA 433.63 USD
77337-644 77358-022 77381-946
(2,4-Dichloro-5-(methylthio)phenyl)methanol ≥97%
(2,4-Dichloro-5-(methylthio)phenyl)methanol

Precaución: for Research Use Only

Formula: C₈H₈Cl₂OS
MW: 223.12 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34764677
CAS Number: 2901084-59-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR