2,4-Dichloro-5-(methylthio)benzaldehyde ≥97%

Supplier: AOBChem USA

Warning

73019-500MG 73019-1G 73019-5G
77335-916EA 367.71 USD
77335-916 77358-020 77381-944
2,4-Dichloro-5-(methylthio)benzaldehyde ≥97%
2,4-Dichloro-5-(methylthio)benzaldehyde

Precaución: for Research Use Only

Formula: C₈H₆Cl₂OS
MW: 221.104 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34764676
CAS Number: 2807450-57-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR