(2,4-Dichloro-5-iodophenyl)(methyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

73018-500MG 73018-1G 73018-5G
77339-454EA 590.19 USD
77339-454 77358-018 77381-942
(2,4-Dichloro-5-iodophenyl)(methyl)sulfane ≥97%
(2,4-Dichloro-5-iodophenyl)(methyl)sulfane

Precaución: for Research Use Only

Formula: C₇H₅Cl₂IS
MW: 318.99 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34764675
CAS Number: 2707564-21-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR