2-Bromo-4-methoxy-6-(methylthio)phenol ≥97%

Supplier: AOBChem USA

Warning

72984-500MG 72984-1G 72984-5G 72984-25G
77333-856EA 148.32 USD
77333-856 77345-286 77369-880 77390-760
2-Bromo-4-methoxy-6-(methylthio)phenol ≥97%
2-Bromo-4-methoxy-6-(methylthio)phenol

Precaución: for Research Use Only

Formula: C₈H₉BrO₂S
MW: 249.125 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34764649
CAS Number: 2091711-80-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR