5-Iodo-2-(p-tolyloxy)pyrimidine ≥95%

Supplier: AOBChem USA

Warning

72971-500MG 72971-1G 72971-5G 72971-25G
77333-854EA 141.11 USD
77333-854 77344-926 77369-878 77390-758
5-Iodo-2-(p-tolyloxy)pyrimidine ≥95%
5-Iodo-2-(p-tolyloxy)pyrimidine

Precaución: for Research Use Only

Formula: C₁₁H₉IN₂O
Storage Temperature: Ambient
MDL Number: MFCD27294611
CAS Number: 1876402-41-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR