5-Bromo-2-(4-methoxyphenoxy)pyrimidine ≥97%

Supplier: AOBChem USA

Warning

72963-1G 72963-5G 72963-25G
77342-134EA 114.33 USD
77342-134 77369-876 77381-938
5-Bromo-2-(4-methoxyphenoxy)pyrimidine ≥97%
5-Bromo-2-(4-methoxyphenoxy)pyrimidine

Precaución: for Research Use Only

Formula: C₁₁H₉BrN₂O₂
MW: 281.105 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03646345
CAS Number: 69033-87-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR