5-Bromo-2-(4-fluorophenoxy)pyrimidine ≥97%

Supplier: AOBChem USA

Warning

72962-500MG 72962-1G 72962-5G 72962-25G
77333-852EA 141.11 USD
77333-852 77344-924 77369-874 77390-756
5-Bromo-2-(4-fluorophenoxy)pyrimidine ≥97%
5-Bromo-2-(4-fluorophenoxy)pyrimidine

Precaución: for Research Use Only

Formula: C₁₀H₆BrFN₂O
Storage Temperature: Ambient
MDL Number: MFCD03646341
CAS Number: 887431-06-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR