5-Bromo-2-(o-tolyloxy)pyrimidine ≥97%

Supplier: AOBChem USA

Warning

72961-1G 72961-5G 72961-25G
77342-036EA 111.24 USD
77342-036 77369-872 77381-936
5-Bromo-2-(o-tolyloxy)pyrimidine ≥97%
5-Bromo-2-(o-tolyloxy)pyrimidine

Precaución: for Research Use Only

Formula: C₁₁H₉BrN₂O
MW: 265.106 g/mol
Storage Temperature: Ambient
CAS Number: 73267-74-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR