Ethyl 4-chloro-6-methoxynicotinate ≥97%

Supplier: AOBChem USA

Warning

72919-500MG 72919-1G 72919-5G 72919-25G
77333-850EA 282.22 USD
77333-850 77358-014 77369-870 77390-754
Ethyl 4-chloro-6-methoxynicotinate ≥97%
Ethyl 4-chloro-6-methoxynicotinate

Precaución: for Research Use Only

Formula: C₉H₁₀ClNO₃
Storage Temperature: Ambient
MDL Number: MFCD18416474
CAS Number: 1261564-03-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR