4-Chloro-2-methoxy-5-(methylthio)pyridine ≥95%

Supplier: AOBChem USA

Warning

72915-500MG 72915-1G 72915-5G 72915-25G
77333-848EA 182.31 USD
77333-848 77347-028 77369-866 77390-752
4-Chloro-2-methoxy-5-(methylthio)pyridine ≥95%
4-Chloro-2-methoxy-5-(methylthio)pyridine

Precaución: for Research Use Only

Formula: C₇H₈ClNOS
MW: 189.663 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34764628
CAS Number: 2807466-17-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR