5-Bromo-3-chloro-2-fluoro-4-methoxybenzoic acid ≥97%

Supplier: AOBChem USA

Warning

72867-500MG 72867-1G 72867-5G 72867-25G
77333-844EA 160.68 USD
77333-844 77345-816 77369-862 77390-748
5-Bromo-3-chloro-2-fluoro-4-methoxybenzoic acid ≥97%
5-Bromo-3-chloro-2-fluoro-4-methoxybenzoic acid

Precaución: for Research Use Only

Formula: C₈H₅BrClFO₃
Storage Temperature: Ambient
MDL Number: MFCD34764610
CAS Number: 2807445-11-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR