2-Chloro-4-isopropoxy-3-methylbenzoic acid ≥97%

Supplier: AOBChem USA

Warning

72847-500MG 72847-1G 72847-5G
77339-718EA 616.97 USD
77339-718 77358-010 77381-932
2-Chloro-4-isopropoxy-3-methylbenzoic acid ≥97%
2-Chloro-4-isopropoxy-3-methylbenzoic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₃ClO₃
MW: 228.672 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34764591
CAS Number: 2807471-82-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR