4-Bromo-1-(tert-butyl)-1H-pyrazole-5-carbaldehyde ≥97%

Supplier: AOBChem USA

Warning

72842-500MG 72842-1G 72842-5G 72842-25G
77333-842EA 122.57 USD
77333-842 77344-010 77369-860 77381-928
4-Bromo-1-(tert-butyl)-1H-pyrazole-5-carbaldehyde ≥97%
4-Bromo-1-(tert-butyl)-1H-pyrazole-5-carbaldehyde

Precaución: for Research Use Only

Formula: C₈H₁₁BrN₂O
MW: 231.09 g/mol
Storage Temperature: Ambient
CAS Number: 1511392-64-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR