4,5-Dibromo-1-(tert-butyl)-1H-pyrazole ≥97%
Supplier: AOBChem USA
|
Warning
|
72841-500MG
72841-1G
72841-5G
77338-352EA
479.98
USD
77338-352
77358-006
77381-926
4,5-Dibromo-1-(tert-butyl)-1H-pyrazole ≥97%
4,5-Dibromo-1-(tert-butyl)-1H-pyrazole
Precaución: for Research Use Only
Formula:
C₇H₁₀Br₂N₂ MW: 281.976 g/mol Storage Temperature: Ambient |
CAS Number:
1895304-42-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...