3,4'-Dichlorobiphenyl (PCB No. 13) ≥97%

Supplier: AOBChem USA

Warning

72823-1G 72823-5G 72823-25G
77341-844EA 105.06 USD
77341-844 77369-854 77381-920
3,4'-Dichlorobiphenyl (PCB No. 13) ≥97%
3,4'-Dichlorobiphenyl (PCB No. 13)

Precaución: for Research Use Only

Formula: C₁₂H₈Cl₂
Storage Temperature: Ambient
MDL Number: MFCD00152402
CAS Number: 2974-90-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR