1-(3-Chloro-4-(methylthio)phenyl)ethanone ≥95%

Supplier: AOBChem USA

Warning

72820-500MG 72820-1G 72820-5G
77336-190EA 380.07 USD
77336-190 77358-004 77381-918
1-(3-Chloro-4-(methylthio)phenyl)ethanone ≥95%
1-(3-Chloro-4-(methylthio)phenyl)ethanone

Precaución: for Research Use Only

Formula: C₉H₉ClOS
MW: 200.685 g/mol
Storage Temperature: Ambient
MDL Number: MFCD17247670
CAS Number: 32467-66-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR