(2-Chloro-1,4-phenylene)bis(methylsulfane) ≥97%

Supplier: AOBChem USA

Warning

72819-500MG 72819-1G 72819-5G 72819-25G
77333-838EA 157.59 USD
77333-838 77345-672 77369-850 77390-744
(2-Chloro-1,4-phenylene)bis(methylsulfane) ≥97%
(2-Chloro-1,4-phenylene)bis(methylsulfane)

Precaución: for Research Use Only

Formula: C₈H₉ClS₂
MW: 204.74 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34764582
CAS Number: 2807464-79-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR