(3-Chloro-5-ethoxy-4-fluorophenyl)(methyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

72818-500MG 72818-1G 72818-5G 72818-25G
77333-836EA 244.11 USD
77333-836 77358-002 77369-848 77390-742
(3-Chloro-5-ethoxy-4-fluorophenyl)(methyl)sulfane ≥97%
(3-Chloro-5-ethoxy-4-fluorophenyl)(methyl)sulfane

Precaución: for Research Use Only

Formula: C₉H₁₀ClFOS
Storage Temperature: Ambient
MDL Number: MFCD34764581
CAS Number: 2711439-91-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR