3-Chloro-5-ethoxy-4-fluorobenzoic acid ≥95%

Supplier: AOBChem USA

Warning

72813-500MG 72813-1G 72813-5G
77333-834EA 347.11 USD
77333-834 77358-000 77381-916
3-Chloro-5-ethoxy-4-fluorobenzoic acid ≥95%
3-Chloro-5-ethoxy-4-fluorobenzoic acid

Precaución: for Research Use Only

Formula: C₉H₈ClFO₃
MW: 218.609 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34764576
CAS Number: 2383623-43-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR